Assay Design
Gene Transcripts
- Gene Symbol
- LCK
- Entrez Gene ID
-
3932
- Gene Name
- LCK proto-oncogene, Src family tyrosine kinase
- Gene Aliases
-
IMD22, LSK, YT16, p56lck, pp58lck
- Chromosome Location
-
Chr.1: 32251239 - 32286167 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC013200.1 |
- |
9 - 10 |
1065 |
76 |
KU178161.1 |
- |
8 - 9 |
965 |
76 |
View Full Product Details