Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198665.1 | NP_001185594.1 | 12 - 13 | 1373 | 96 |
NM_001301084.1 | NP_001288013.1 | 13 - 14 | 1256 | 96 | |
NM_015313.2 | NP_056128.1 | 13 - 14 | 1430 | 96 | |
XM_006718805.3 | XP_006718868.1 | 12 - 13 | 1714 | 96 | |
XM_011542720.2 | XP_011541022.1 | 13 - 14 | 1371 | 96 | |
XM_017017420.1 | XP_016872909.1 | 11 - 12 | 1886 | 96 | |
XM_017017421.1 | XP_016872910.1 | 13 - 14 | 1646 | 96 | |
XM_017017422.1 | XP_016872911.1 | 12 - 13 | 1138 | 96 | |
GenBank mRNA | AB002380.2 | - | 8 - 9 | 792 | 96 |
AF180681.1 | - | 13 - 14 | 1102 | 96 | |
AK294803.1 | - | 13 - 14 | 1231 | 96 | |
AK304316.1 | - | 13 - 14 | 1654 | 96 | |
BC063117.1 | - | 12 - 13 | 1376 | 96 | |
BX647785.1 | - | 13 - 14 | 1232 | 96 | |
BX648515.1 | - | 13 - 14 | 1237 | 96 |