Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077196.1 | NP_001070664.1 | 14 - 15 | 1311 | 74 |
NM_001077197.1 | NP_001070665.1 | 18 - 19 | 1841 | 74 | |
NM_001077358.1 | NP_001070826.1 | 16 - 17 | 1551 | 74 | |
NM_016953.3 | NP_058649.3 | 17 - 18 | 2810 | 74 | |
GenBank mRNA | AB036704.1 | - | 17 - 18 | 2810 | 74 |
AB038041.1 | - | 18 - 19 | 1841 | 74 | |
AF281865.1 | - | 16 - 17 | 1523 | 74 | |
AJ251509.1 | - | 14 - 15 | 1310 | 74 | |
AJ278682.1 | - | 18 - 19 | 1884 | 74 | |
AK074273.1 | - | 7 - 8 | 708 | 74 | |
AK294076.1 | - | 2 - 3 | 99 | 74 | |
BC112393.1 | - | 17 - 18 | 2801 | 74 | |
BC114431.1 | - | 17 - 18 | 2801 | 74 | |
HM771389.1 | - | 17 - 18 | 2492 | 74 | |
HM771390.1 | - | 17 - 18 | 2492 | 74 |