Assay Design



Gene Transcripts

Gene Symbol
C2orf16
Entrez Gene ID
84226
Gene Name
chromosome 2 open reading frame 16
Gene Aliases
-
Chromosome Location
Chr.2: 27576522 - 27582722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032266.3 NP_115642.3 1 - 1 6150 114
GenBank mRNA AL136898.1 - 1 - 1 3010 114
CR749335.1 - 1 - 1 6150 114

View Full Product Details