Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190481.1 | NP_001177410.1 | 4 - 5 | 817 | 155 |
NM_022111.3 | NP_071394.2 | 4 - 5 | 817 | 155 | |
XM_011541938.2 | XP_011540240.1 | 4 - 5 | 840 | 155 | |
XM_011541939.2 | XP_011540241.1 | 4 - 5 | 834 | 155 | |
XM_011541940.2 | XP_011540242.1 | 4 - 5 | 834 | 155 | |
GenBank mRNA | AF297866.1 | - | 4 - 5 | 817 | 155 |
AK308193.1 | - | 4 - 5 | 846 | 155 | |
BC018670.1 | - | 4 - 5 | 837 | 155 | |
BC038991.2 | - | 4 - 5 | 843 | 155 | |
BC113116.1 | - | 4 - 5 | 810 | 155 | |
BC115025.1 | - | 4 - 5 | 810 | 155 | |
BC115026.1 | - | 4 - 5 | 810 | 155 | |
BC137279.1 | - | 4 - 5 | 796 | 155 | |
BC140789.1 | - | 4 - 5 | 796 | 155 |