Assay Design



Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
Chromosome Location
Chr.1: 35720465 - 35769985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190481.1 NP_001177410.1 5 - 6 904 100
NM_022111.3 NP_071394.2 5 - 6 904 100
XM_011541938.2 XP_011540240.1 5 - 6 927 100
XM_011541939.2 XP_011540241.1 5 - 6 921 100
XM_011541940.2 XP_011540242.1 5 - 6 921 100
XM_011541941.2 XP_011540243.1 4 - 5 759 100
GenBank mRNA AF297866.1 - 5 - 6 904 100
AK308193.1 - 5 - 6 933 100
BC018670.1 - 5 - 6 924 100
BC038991.2 - 5 - 6 930 100
BC113116.1 - 5 - 6 897 100
BC115025.1 - 5 - 6 897 100
BC115026.1 - 5 - 6 897 100
BC137279.1 - 5 - 6 883 100
BC140789.1 - 5 - 6 883 100

View Full Product Details