Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005469.3 | NP_005460.2 | 5 - 6 | 934 | 73 |
XM_005260239.3 | XP_005260296.1 | 4 - 5 | 796 | 73 | |
XM_011528480.2 | XP_011526782.1 | 6 - 7 | 958 | 73 | |
GenBank mRNA | AF014404.1 | - | 5 - 6 | 870 | 73 |
AF124264.1 | - | 5 - 6 | 840 | 73 | |
BC117155.1 | - | 5 - 6 | 862 | 73 | |
BC117157.1 | - | 5 - 6 | 862 | 73 | |
BM986979.1 | - | 1 - 2 | 255 | 73 | |
CR541771.1 | - | 5 - 6 | 840 | 73 | |
CR541778.1 | - | 5 - 6 | 840 | 73 |