Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099436.3 | NP_001092906.3 | 5 - 6 | 717 | 159 |
NM_001284364.2 | NP_001271293.2 | 5 - 6 | 717 | 159 | |
NM_001284365.2 | NP_001271294.1 | 5 - 6 | 640 | 159 | |
XM_005254289.2 | XP_005254346.1 | 6 - 7 | 732 | 159 | |
XM_017022068.1 | XP_016877557.1 | 6 - 7 | 766 | 159 | |
XM_017022069.1 | XP_016877558.1 | 6 - 7 | 770 | 159 | |
XM_017022070.1 | XP_016877559.1 | 6 - 7 | 783 | 159 | |
XM_017022071.1 | XP_016877560.1 | 6 - 7 | 770 | 159 | |
XM_017022072.1 | XP_016877561.1 | 5 - 6 | 946 | 159 | |
GenBank mRNA | AK293181.1 | - | 5 - 6 | 570 | 159 |
AK294241.1 | - | 5 - 6 | 651 | 159 | |
AK294245.1 | - | 5 - 6 | 647 | 159 | |
AK294251.1 | - | 5 - 6 | 651 | 159 | |
AK298879.1 | - | 5 - 6 | 574 | 159 | |
AK299380.1 | - | 5 - 6 | 644 | 159 | |
BC157884.1 | - | 5 - 6 | 699 | 159 | |
BQ927030.1 | - | 5 - 6 | 605 | 159 |