Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002305.3 | NP_002296.1 | 2 - 3 | 189 | 80 |
GenBank mRNA | AB097036.1 | - | 2 - 3 | 141 | 80 |
AK312161.1 | - | 2 - 3 | 159 | 80 | |
AL832117.1 | - | 2 - 3 | 159 | 80 | |
BC001693.1 | - | 2 - 3 | 142 | 80 | |
BC020675.1 | - | 2 - 3 | 159 | 80 | |
BT006775.1 | - | 1 - 2 | 92 | 80 | |
CR456511.1 | - | 2 - 3 | 189 | 80 | |
EU363770.1 | - | 2 - 3 | 161 | 80 | |
J04456.1 | - | 2 - 3 | 141 | 80 | |
X14829.1 | - | 2 - 3 | 99 | 80 | |
X15256.1 | - | 2 - 3 | 141 | 80 |