Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006498.2 | NP_006489.1 | 1 - 2 | 121 | 61 |
GenBank mRNA | BC059782.1 | - | 1 - 2 | 47 | 61 |
CD701729.1 | - | 1 - 2 | 195 | 61 | |
CR456512.1 | - | 1 - 2 | 161 | 61 | |
M87842.1 | - | 1 - 2 | 35 | 61 |