Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000575.4 | NP_000566.3 | 4 - 5 | 1284 | 75 |
GenBank mRNA | AK314850.1 | - | 4 - 5 | 379 | 75 |
BC013142.2 | - | 3 - 4 | 329 | 75 | |
BT007014.1 | - | 3 - 4 | 320 | 75 | |
CR457414.1 | - | 3 - 4 | 320 | 75 | |
M15329.1 | - | 3 - 4 | 325 | 75 | |
X02531.1 | - | 3 - 4 | 320 | 75 | |
X02851.1 | - | 4 - 5 | 378 | 75 | |
X56086.1 | - | 4 - 5 | 346 | 75 |