Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 13 - 14 | 1764 | 79 |
NM_001010932.2 | NP_001010932.1 | 13 - 14 | 1749 | 79 | |
XM_006715956.2 | XP_006716019.1 | 14 - 15 | 1727 | 79 | |
XM_011516115.2 | XP_011514417.1 | 14 - 15 | 1712 | 79 | |
GenBank mRNA | AK291674.1 | - | 13 - 14 | 1699 | 79 |
AK313694.1 | - | 13 - 14 | 1599 | 79 | |
BC130284.1 | - | 13 - 14 | 1612 | 79 | |
BC130286.1 | - | 13 - 14 | 1612 | 79 | |
M29145.1 | - | 13 - 14 | 1638 | 79 | |
M55379.1 | - | 13 - 14 | 1522 | 79 | |
M60718.1 | - | 13 - 14 | 1609 | 79 | |
M73239.1 | - | 13 - 14 | 1602 | 79 | |
M73240.1 | - | 13 - 14 | 1587 | 79 |