Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 17 - 18 | 2238 | 98 |
NM_001010932.2 | NP_001010932.1 | 17 - 18 | 2223 | 98 | |
XM_006715956.2 | XP_006716019.1 | 18 - 19 | 2201 | 98 | |
XM_011516115.2 | XP_011514417.1 | 18 - 19 | 2186 | 98 | |
GenBank mRNA | AK291674.1 | - | 17 - 18 | 2173 | 98 |
AK313694.1 | - | 17 - 18 | 2073 | 98 | |
BC130284.1 | - | 17 - 18 | 2086 | 98 | |
BC130286.1 | - | 17 - 18 | 2086 | 98 | |
M29145.1 | - | 17 - 18 | 2112 | 98 | |
M55379.1 | - | 17 - 18 | 1996 | 98 | |
M60718.1 | - | 17 - 18 | 2083 | 98 | |
M73239.1 | - | 17 - 18 | 2076 | 98 | |
M73240.1 | - | 17 - 18 | 2061 | 98 | |
X16323.1 | - | 17 - 18 | 2145 | 98 |