Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 5 - 6 | 856 | 70 |
NM_001010931.2 | NP_001010931.1 | 5 - 6 | 856 | 70 | |
NM_001010932.2 | NP_001010932.1 | 5 - 6 | 841 | 70 | |
NM_001010933.2 | NP_001010933.1 | 5 - 6 | 841 | 70 | |
XM_006715956.2 | XP_006716019.1 | 6 - 7 | 819 | 70 | |
XM_011516115.2 | XP_011514417.1 | 6 - 7 | 804 | 70 | |
XM_017012097.1 | XP_016867586.1 | 6 - 7 | 819 | 70 | |
XM_017012098.1 | XP_016867587.1 | 6 - 7 | 804 | 70 | |
GenBank mRNA | AK291674.1 | - | 5 - 6 | 791 | 70 |
AK313694.1 | - | 5 - 6 | 691 | 70 | |
BC022308.1 | - | 5 - 6 | 690 | 70 | |
BC130284.1 | - | 5 - 6 | 704 | 70 | |
BC130286.1 | - | 5 - 6 | 704 | 70 | |
BM971901.1 | - | 3 - 4 | 70 | ||
L02931.1 | - | 5 - 6 | 689 | 70 | |
M29145.1 | - | 5 - 6 | 730 | 70 | |
M55379.1 | - | 5 - 6 | 614 | 70 | |
M60718.1 | - | 5 - 6 | 701 | 70 | |
M73239.1 | - | 5 - 6 | 694 | 70 | |
M73240.1 | - | 5 - 6 | 679 | 70 | |
M77227.1 | - | 5 - 6 | 711 | 70 | |
X16323.1 | - | 5 - 6 | 763 | 70 | |
X57574.1 | - | 5 - 6 | 706 | 70 |