Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 6 - 7 | 969 | 85 |
NM_001010931.2 | NP_001010931.1 | 6 - 7 | 969 | 85 | |
NM_001010932.2 | NP_001010932.1 | 6 - 7 | 954 | 85 | |
NM_001010933.2 | NP_001010933.1 | 6 - 7 | 954 | 85 | |
XM_006715956.2 | XP_006716019.1 | 7 - 8 | 932 | 85 | |
XM_011516115.2 | XP_011514417.1 | 7 - 8 | 917 | 85 | |
XM_017012097.1 | XP_016867586.1 | 7 - 8 | 932 | 85 | |
XM_017012098.1 | XP_016867587.1 | 7 - 8 | 917 | 85 | |
GenBank mRNA | AK291674.1 | - | 6 - 7 | 904 | 85 |
AK313694.1 | - | 6 - 7 | 804 | 85 | |
BC022308.1 | - | 6 - 7 | 803 | 85 | |
BC130284.1 | - | 6 - 7 | 817 | 85 | |
BC130286.1 | - | 6 - 7 | 817 | 85 | |
BM971901.1 | - | 2 - 3 | 85 | ||
L02931.1 | - | 6 - 7 | 802 | 85 | |
M29145.1 | - | 6 - 7 | 843 | 85 | |
M55379.1 | - | 6 - 7 | 727 | 85 | |
M60718.1 | - | 6 - 7 | 814 | 85 | |
M73239.1 | - | 6 - 7 | 807 | 85 | |
M73240.1 | - | 6 - 7 | 792 | 85 | |
M77227.1 | - | 6 - 7 | 824 | 85 | |
X16323.1 | - | 6 - 7 | 876 | 85 | |
X57574.1 | - | 6 - 7 | 819 | 85 |