Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003184.3 | NP_003175.1 | 3 - 4 | 567 | 72 |
XM_006716621.3 | XP_006716684.1 | 3 - 4 | 591 | 72 | |
XM_011517259.2 | XP_011515561.1 | 3 - 4 | 591 | 72 | |
XM_017013779.1 | XP_016869268.1 | 3 - 4 | 591 | 72 | |
XM_017013783.1 | XP_016869272.1 | 2 - 3 | 204 | 72 | |
XM_017013788.1 | XP_016869277.1 | 2 - 3 | 204 | 72 | |
XM_017013789.1 | XP_016869278.1 | 2 - 3 | 204 | 72 | |
XM_017013790.1 | XP_016869279.1 | 2 - 3 | 204 | 72 | |
GenBank mRNA | AF026445.1 | - | 4 - 5 | 562 | 72 |
AF040701.1 | - | 3 - 4 | 297 | 72 | |
AF057694.1 | - | 3 - 4 | 354 | 72 | |
AK316592.1 | - | 3 - 4 | 596 | 72 | |
BC035673.2 | - | 3 - 4 | 556 | 72 | |
BC047732.1 | - | 3 - 4 | 563 | 72 | |
BC064830.1 | - | 3 - 4 | 571 | 72 |