Assay Design



Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 16 - 17 3402 74
XM_005267064.3 XP_005267121.1 15 - 16 5481 74
XM_005267065.3 XP_005267122.1 17 - 18 3428 74
XM_005267066.2 XP_005267123.1 15 - 16 4285 74
XM_006715521.1 XP_006715584.1 17 - 18 3655 74
XM_006715522.3 XP_006715585.1 15 - 16 5522 74
XM_011535981.2 XP_011534283.1 15 - 16 6278 74
XM_011535982.2 XP_011534284.1 14 - 15 6239 74
XM_011535983.2 XP_011534285.1 16 - 17 4703 74
XM_017011102.1 XP_016866591.1 14 - 15 4248 74
GenBank mRNA AB033035.1 - 9 - 10 2275 74
BC089428.1 - 11 - 12 2513 74
BC140864.1 - 14 - 15 3139 74
BC151134.1 - 14 - 15 3139 74

View Full Product Details