Assay Design



Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 6 - 7 942 113
XM_005267064.3 XP_005267121.1 5 - 6 3021 113
XM_005267065.3 XP_005267122.1 7 - 8 968 113
XM_005267066.2 XP_005267123.1 5 - 6 1825 113
XM_006715521.1 XP_006715584.1 7 - 8 1195 113
XM_006715522.3 XP_006715585.1 5 - 6 3062 113
XM_011535981.2 XP_011534283.1 5 - 6 3818 113
XM_011535982.2 XP_011534284.1 5 - 6 3818 113
XM_011535983.2 XP_011534285.1 6 - 7 2243 113
XM_017011102.1 XP_016866591.1 5 - 6 1827 113
GenBank mRNA AJ420468.1 - 3 - 4 180 113
AK056300.1 - 5 - 6 841 113
AK294798.1 - 5 - 6 996 113
AK307631.1 - 5 - 6 895 113
BC140864.1 - 4 - 5 679 113
BC151134.1 - 4 - 5 679 113

View Full Product Details