Assay Design



Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 7 - 8 1093 87
XM_005267064.3 XP_005267121.1 6 - 7 3172 87
XM_005267065.3 XP_005267122.1 8 - 9 1119 87
XM_005267066.2 XP_005267123.1 6 - 7 1976 87
XM_006715521.1 XP_006715584.1 8 - 9 1346 87
XM_006715522.3 XP_006715585.1 6 - 7 3213 87
XM_011535981.2 XP_011534283.1 6 - 7 3969 87
XM_011535982.2 XP_011534284.1 6 - 7 3969 87
XM_011535983.2 XP_011534285.1 7 - 8 2394 87
XM_017011102.1 XP_016866591.1 6 - 7 1978 87
GenBank mRNA AJ420468.1 - 4 - 5 331 87
AK056300.1 - 6 - 7 992 87
AK293288.1 - 6 - 7 1024 87
AK294798.1 - 6 - 7 1147 87
AK307631.1 - 6 - 7 1046 87
BC089428.1 - 2 - 3 204 87
BC140864.1 - 5 - 6 830 87
BC151134.1 - 5 - 6 830 87

View Full Product Details