Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023034.1 | NP_075447.1 | 14 - 15 | 3136 | 123 |
GenBank mRNA | AF332469.1 | - | 14 - 15 | 3136 | 123 |
AJ295990.1 | - | 14 - 15 | 2931 | 123 | |
BC101717.1 | - | 14 - 15 | 2710 | 123 | |
BC113469.1 | - | 14 - 15 | 2710 | 123 | |
BC115006.1 | - | 14 - 15 | 2731 | 123 | |
BC143510.1 | - | 14 - 15 | 2710 | 123 |