Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281482.1 | NP_001268411.1 | 5 - 6 | 580 | 97 |
NM_001281483.1 | NP_001268412.1 | 4 - 5 | 470 | 97 | |
NM_018209.3 | NP_060679.1 | 5 - 6 | 580 | 97 | |
NM_175609.2 | NP_783202.1 | 5 - 6 | 580 | 97 | |
XM_006723823.2 | XP_006723886.1 | 5 - 6 | 563 | 97 | |
XM_011528901.1 | XP_011527203.1 | 5 - 6 | 563 | 97 | |
XM_017027950.1 | XP_016883439.1 | 5 - 6 | 646 | 97 | |
GenBank mRNA | AK001629.1 | - | 5 - 6 | 552 | 97 |
AK027268.1 | - | 2 - 3 | 272 | 97 | |
AK223427.1 | - | 5 - 6 | 501 | 97 | |
AK296351.1 | - | 4 - 5 | 442 | 97 | |
AK308161.1 | - | 5 - 6 | 563 | 97 | |
BC000786.1 | - | 5 - 6 | 540 | 97 | |
BC006085.1 | - | 5 - 6 | 531 | 97 | |
BC011876.1 | - | 5 - 6 | 540 | 97 | |
BC028233.1 | - | 5 - 6 | 500 | 97 |