Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130420.1 | NP_001123892.1 | 4 - 5 | 507 | 141 |
NM_003075.3 | NP_003066.2 | 4 - 5 | 507 | 141 | |
NM_139067.2 | NP_620706.1 | 4 - 5 | 507 | 141 | |
XM_005269101.1 | XP_005269158.1 | 4 - 5 | 444 | 141 | |
XM_005269102.1 | XP_005269159.1 | 4 - 5 | 444 | 141 | |
XM_005269103.1 | XP_005269160.1 | 4 - 5 | 444 | 141 | |
XM_005269104.1 | XP_005269161.1 | 4 - 5 | 444 | 141 | |
XM_011538693.2 | XP_011536995.1 | 4 - 5 | 423 | 141 | |
XM_017019884.1 | XP_016875373.1 | 4 - 5 | 444 | 141 | |
XM_017019885.1 | XP_016875374.1 | 4 - 5 | 444 | 141 | |
XM_017019886.1 | XP_016875375.1 | 4 - 5 | 444 | 141 | |
XM_017019887.1 | XP_016875376.1 | 4 - 5 | 401 | 141 | |
GenBank mRNA | AB209006.1 | - | 4 - 5 | 507 | 141 |
AB209293.1 | - | 4 - 5 | 416 | 141 | |
AK293896.1 | - | 4 - 5 | 394 | 141 | |
AK315153.1 | - | 4 - 5 | 407 | 141 | |
BC013045.1 | - | 4 - 5 | 433 | 141 | |
BC026222.2 | - | 4 - 5 | 426 | 141 | |
BT009924.1 | - | 4 - 5 | 401 | 141 | |
U66616.1 | - | 4 - 5 | 423 | 141 |