Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135195.1 | NP_001128667.1 | 6 - 7 | 1177 | 90 |
NM_173596.2 | NP_775867.2 | 8 - 9 | 1206 | 90 | |
XM_005268803.1 | XP_005268860.1 | 6 - 7 | 1094 | 90 | |
XM_011538198.1 | XP_011536500.1 | 6 - 7 | 1159 | 90 | |
XM_011538199.1 | XP_011536501.1 | 8 - 9 | 1213 | 90 | |
XM_011538200.1 | XP_011536502.1 | 7 - 8 | 1163 | 90 | |
XM_011538201.1 | XP_011536503.1 | 7 - 8 | 1137 | 90 | |
XM_017019185.1 | XP_016874674.1 | 6 - 7 | 1137 | 90 | |
GenBank mRNA | AK172768.1 | - | 6 - 7 | 1140 | 90 |
AK313188.1 | - | 8 - 9 | 1169 | 90 | |
BC027884.1 | - | 6 - 7 | 1051 | 90 |