Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022351.4 | NP_071746.1 | 9 - 10 | 943 | 96 |
XM_011517213.2 | XP_011515515.1 | 9 - 10 | 868 | 96 | |
GenBank mRNA | AF193756.1 | - | 5 - 6 | 461 | 96 |
AF414126.2 | - | 10 - 11 | 1032 | 96 | |
AK095533.1 | - | 9 - 10 | 856 | 96 | |
AY224210.1 | - | 9 - 10 | 749 | 96 | |
BC068448.1 | - | 9 - 10 | 856 | 96 |