Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317934.1 | NP_001304863.1 | 3 - 4 | 764 | 64 |
NM_001317935.1 | NP_001304864.1 | 3 - 4 | 752 | 64 | |
NM_020894.3 | NP_065945.2 | 3 - 4 | 865 | 64 | |
XM_017008490.1 | XP_016863979.1 | 3 - 4 | 586 | 64 | |
XM_017008491.1 | XP_016863980.1 | 3 - 4 | 678 | 64 | |
XM_017008492.1 | XP_016863981.1 | 3 - 4 | 676 | 64 | |
XM_017008493.1 | XP_016863982.1 | 3 - 4 | 552 | 64 | |
XM_017008494.1 | XP_016863983.1 | 3 - 4 | 437 | 64 | |
XM_017008496.1 | XP_016863985.1 | 3 - 4 | 1189 | 64 | |
XM_017008497.1 | XP_016863986.1 | 3 - 4 | 1188 | 64 | |
XM_017008498.1 | XP_016863987.1 | 2 - 3 | 420 | 64 | |
XM_017008499.1 | XP_016863988.1 | 3 - 4 | 693 | 64 | |
GenBank mRNA | AB040963.1 | - | 3 - 4 | 865 | 64 |
BC110331.1 | - | 3 - 4 | 764 | 64 | |
BC140901.1 | - | 3 - 4 | 602 | 64 |