Assay Design



Gene Transcripts

Gene Symbol
OR51I1
Entrez Gene ID
390063
Gene Name
olfactory receptor family 51 subfamily I member 1
Gene Aliases
-
Chromosome Location
Chr.11: 5440542 - 5441553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005288.2 NP_001005288.1 1 - 1 905 121
GenBank mRNA BC151148.1 - 1 - 1 905 121

View Full Product Details