Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182488.3 | NP_872294.2 | 1 - 2 | 306 | 63 |
XM_005266282.2 | XP_005266339.1 | 1 - 2 | 220 | 63 | |
XM_017020416.1 | XP_016875905.1 | 1 - 2 | 191 | 63 | |
GenBank mRNA | AF022789.1 | - | 1 - 2 | 156 | 63 |
AK289685.1 | - | 1 - 2 | 220 | 63 | |
BC026072.1 | - | 1 - 2 | 307 | 63 |