Assay Design



Gene Transcripts

Gene Symbol
PHYHIP
Entrez Gene ID
9796
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein
Gene Aliases
DYRK1AP3, PAHX-AP, PAHXAP1
Chromosome Location
Chr.8: 22219703 - 22232338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099335.1 NP_001092805.1 2 - 3 667 84
GenBank mRNA D87463.1 - 2 - 3 376 84

View Full Product Details