Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242846.1 | NP_001229775.1 | 1 - 2 | 89 | 82 |
NM_001242848.1 | NP_001229777.1 | 1 - 2 | 89 | 82 | |
NM_001242850.1 | NP_001229779.1 | 1 - 2 | 89 | 82 | |
NM_030963.3 | NP_112225.2 | 1 - 2 | 89 | 82 | |
XM_011536161.2 | XP_011534463.1 | 1 - 2 | 587 | 82 | |
XM_011536162.2 | XP_011534464.1 | 1 - 2 | 587 | 82 | |
XM_011536163.2 | XP_011534465.1 | 1 - 2 | 587 | 82 | |
XM_011536164.2 | XP_011534466.1 | 1 - 2 | 587 | 82 | |
XM_017011338.1 | XP_016866827.1 | 1 - 2 | 587 | 82 | |
GenBank mRNA | AK027558.1 | - | 1 - 2 | 78 | 82 |
AL136829.1 | - | 1 - 2 | 52 | 82 | |
BG708628.1 | - | 1 - 2 | 324 | 82 | |
DA145911.1 | - | 1 - 2 | 76 | 82 | |
DB267873.1 | - | 1 - 2 | 91 | 82 |