Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256796.1 | NP_001243725.1 | 10 - 11 | 1330 | 79 |
NM_001261397.1 | NP_001248326.1 | 9 - 10 | 1201 | 79 | |
NM_002560.2 | NP_002551.2 | 9 - 10 | 1282 | 79 | |
NR_046372.1 | 10 - 11 | 1320 | 79 | ||
NR_046373.1 | 8 - 9 | 1134 | 79 | ||
XM_011538416.2 | XP_011536718.1 | 5 - 6 | 645 | 79 | |
GenBank mRNA | AF000234.1 | - | 9 - 10 | 878 | 79 |
AK293599.1 | - | 6 - 7 | 772 | 79 | |
AK293660.1 | - | 8 - 9 | 917 | 79 | |
AK297704.1 | - | 8 - 9 | 866 | 79 | |
AK301080.1 | - | 9 - 10 | 966 | 79 | |
AK308549.1 | - | 10 - 11 | 1052 | 79 | |
AK308868.1 | - | 9 - 10 | 1973 | 79 | |
BC017458.1 | - | 9 - 10 | 1011 | 79 | |
BC033826.1 | - | 9 - 10 | 1002 | 79 | |
BT019738.1 | - | 9 - 10 | 978 | 79 | |
BT019739.1 | - | 9 - 10 | 897 | 79 | |
JF807485.1 | - | 2 - 3 | 135 | 79 | |
JX471561.1 | - | 2 - 3 | 174 | 79 | |
U83993.1 | - | 9 - 10 | 1287 | 79 | |
Y07684.1 | - | 9 - 10 | 1005 | 79 |