Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260474.1 | NP_001247403.1 | 2 - 3 | 243 | 64 |
NM_001260475.1 | NP_001247404.1 | 2 - 3 | 243 | 64 | |
NM_001260476.1 | NP_001247405.1 | 2 - 3 | 143 | 64 | |
NM_001260477.1 | NP_001247406.1 | 2 - 3 | 350 | 64 | |
NM_018669.5 | NP_061139.2 | 2 - 3 | 243 | 64 | |
NM_033661.4 | NP_387510.1 | 2 - 3 | 243 | 64 | |
NR_048535.1 | 2 - 3 | 135 | 64 | ||
XM_011529433.1 | XP_011527735.1 | 2 - 3 | 506 | 64 | |
XM_017028262.1 | XP_016883751.1 | 2 - 3 | 411 | 64 | |
XM_017028263.1 | XP_016883752.1 | 2 - 3 | 170 | 64 | |
XM_017028264.1 | XP_016883753.1 | 2 - 3 | 170 | 64 | |
GenBank mRNA | AJ243912.1 | - | 2 - 3 | 187 | 64 |
AJ243913.1 | - | 2 - 3 | 187 | 64 | |
AK056343.1 | - | 2 - 3 | 350 | 64 | |
AK292923.1 | - | 2 - 3 | 184 | 64 | |
AK298015.1 | - | 2 - 3 | 135 | 64 | |
AK315008.1 | - | 2 - 3 | 186 | 64 | |
BC001074.2 | - | 2 - 3 | 122 | 64 | |
BC006341.2 | - | 2 - 3 | 191 | 64 | |
BG722541.1 | - | 2 - 3 | 217 | 64 | |
BU541429.1 | - | 2 - 3 | 169 | 64 | |
DC303088.1 | - | 2 - 3 | 243 | 64 |