Assay Design
Gene Transcripts
- Gene Symbol
- KRTAP4-2
- Entrez Gene ID
-
85291
- Gene Name
- keratin associated protein 4-2
- Gene Aliases
-
KAP4.2, KRTAP4.2
- Chromosome Location
-
Chr.17: 41177446 - 41178208 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_033062.3 |
NP_149051.1 |
1 - 1 |
646 |
95 |
GenBank mRNA |
AJ406934.1 |
- |
1 - 1 |
646 |
95 |
View Full Product Details