Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159596.1 | NP_001153068.1 | 3 - 4 | 698 | 138 |
NM_017693.3 | NP_060163.2 | 5 - 6 | 1382 | 138 | |
GenBank mRNA | AF339793.1 | - | 1 - 2 | 771 | 138 |
AF411385.1 | - | 5 - 6 | 1382 | 138 | |
AK000166.1 | - | 3 - 4 | 594 | 138 | |
AK225229.1 | - | 3 - 4 | 594 | 138 | |
AK290658.1 | - | 5 - 6 | 1251 | 138 | |
AK304391.1 | - | 4 - 5 | 1136 | 138 | |
BC020663.1 | - | 3 - 4 | 526 | 138 | |
BC051813.2 | - | 3 - 4 | 507 | 138 | |
BC075084.2 | - | 3 - 4 | 730 | 138 | |
BC075085.2 | - | 3 - 4 | 730 | 138 | |
BC112339.1 | - | 3 - 4 | 453 | 138 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 3 - 4 | 825 | 138 |
GenBank mRNA | AB209312.1 | - | 3 - 4 | 617 | 138 |
LM994822.1 | - | 3 - 4 | 703 | 138 |