Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159596.1 | NP_001153068.1 | 4 - 5 | 807 | 111 |
NM_017693.3 | NP_060163.2 | 6 - 7 | 1491 | 111 | |
GenBank mRNA | AF339793.1 | - | 2 - 3 | 880 | 111 |
AF411385.1 | - | 6 - 7 | 1491 | 111 | |
AK000166.1 | - | 4 - 5 | 703 | 111 | |
AK225229.1 | - | 4 - 5 | 703 | 111 | |
AK290658.1 | - | 6 - 7 | 1360 | 111 | |
AK304391.1 | - | 5 - 6 | 1245 | 111 | |
BC020663.1 | - | 4 - 5 | 635 | 111 | |
BC051813.2 | - | 4 - 5 | 616 | 111 | |
BC075084.2 | - | 4 - 5 | 839 | 111 | |
BC075085.2 | - | 4 - 5 | 839 | 111 | |
BC112339.1 | - | 4 - 5 | 562 | 111 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 4 - 5 | 934 | 111 |
GenBank mRNA | AB209312.1 | - | 4 - 5 | 726 | 111 |
LM994822.1 | - | 4 - 5 | 812 | 111 |