Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159596.1 | NP_001153068.1 | 7 - 8 | 1120 | 90 |
NM_017693.3 | NP_060163.2 | 9 - 10 | 1804 | 90 | |
GenBank mRNA | AF339783.1 | - | 4 - 5 | 395 | 90 |
AF339793.1 | - | 5 - 6 | 1193 | 90 | |
AF411385.1 | - | 9 - 10 | 1804 | 90 | |
AK000166.1 | - | 7 - 8 | 1016 | 90 | |
AK225229.1 | - | 7 - 8 | 1016 | 90 | |
AK290658.1 | - | 9 - 10 | 1673 | 90 | |
AK304391.1 | - | 7 - 8 | 1425 | 90 | |
BC020663.1 | - | 7 - 8 | 948 | 90 | |
BC051813.2 | - | 7 - 8 | 929 | 90 | |
BC075084.2 | - | 7 - 8 | 1152 | 90 | |
BC075085.2 | - | 7 - 8 | 1152 | 90 | |
BC112339.1 | - | 7 - 8 | 875 | 90 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 7 - 8 | 1247 | 90 |
GenBank mRNA | AB209312.1 | - | 7 - 8 | 1039 | 90 |
LM994822.1 | - | 7 - 8 | 1125 | 90 |