Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006573.4 | NP_006564.1 | 3 - 4 | 753 | 73 |
GenBank mRNA | AF116456.1 | - | 3 - 4 | 604 | 73 |
AF132600.1 | - | 3 - 4 | 622 | 73 | |
AF134715.1 | - | 3 - 4 | 630 | 73 | |
AF136293.1 | - | 3 - 4 | 486 | 73 | |
AF186114.1 | - | 3 - 4 | 643 | 73 | |
AY129225.1 | - | 3 - 4 | 486 | 73 | |
AY129226.1 | - | 3 - 4 | 255 | 73 | |
AY129227.1 | - | 2 - 3 | 153 | 73 | |
AY129228.1 | - | 2 - 3 | 105 | 73 | |
AY358881.1 | - | 3 - 4 | 655 | 73 | |
BC020674.1 | - | 3 - 4 | 665 | 73 | |
CR541818.1 | - | 3 - 4 | 486 | 73 |