Assay Design



Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 18 - 19 3087 80
NM_004592.3 NP_004583.2 17 - 18 2931 80
XM_011538653.1 XP_011536955.1 17 - 18 2853 80
XM_011538654.2 XP_011536956.1 18 - 19 3039 80
XM_017019798.1 XP_016875287.1 16 - 17 2697 80
XM_017019799.1 XP_016875288.1 18 - 19 3526 80
GenBank mRNA AA410512.1 - 1 - 2 80
BC136678.1 - 17 - 18 2881 80
BC144364.1 - 18 - 19 3037 80
U08377.1 - 17 - 18 2908 80

View Full Product Details