Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000562.2 | NP_000553.1 | 7 - 8 | 1222 | 102 |
GenBank mRNA | BC020702.1 | - | 7 - 8 | 1195 | 102 |
BC132911.1 | - | 7 - 8 | 1114 | 102 | |
BC132913.1 | - | 7 - 8 | 1114 | 102 | |
M16974.1 | - | 7 - 8 | 1227 | 102 |