Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018963.4 | NP_061836.2 | 27 - 28 | 3503 | 88 |
NM_033656.3 | NP_387505.1 | 27 - 28 | 3503 | 88 | |
XM_011529611.1 | XP_011527913.1 | 27 - 28 | 3536 | 88 | |
XM_011529612.2 | XP_011527914.1 | 20 - 21 | 2399 | 88 | |
XM_011529613.1 | XP_011527915.1 | 5 - 6 | 498 | 88 | |
XM_017028373.1 | XP_016883862.1 | 28 - 29 | 3308 | 88 | |
XM_017028374.1 | XP_016883863.1 | 3 - 4 | 2138 | 88 | |
GenBank mRNA | AB080586.1 | - | 27 - 28 | 3307 | 88 |
AB080587.1 | - | 27 - 28 | 3307 | 88 | |
AJ238214.2 | - | 27 - 28 | 3307 | 88 | |
AJ292465.1 | - | 27 - 28 | 3287 | 88 | |
AJ292466.1 | - | 27 - 28 | 3287 | 88 | |
AK125906.1 | - | 12 - 13 | 1600 | 88 |