Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 13 - 14 | 1858 | 71 |
XM_005249549.1 | XP_005249606.1 | 10 - 11 | 1450 | 71 | |
XM_006716041.2 | XP_006716104.1 | 13 - 14 | 1882 | 71 | |
XM_011516367.1 | XP_011514669.1 | 13 - 14 | 1724 | 71 | |
XM_011516368.2 | XP_011514670.1 | 13 - 14 | 1882 | 71 | |
XM_011516369.2 | XP_011514671.1 | 13 - 14 | 1713 | 71 | |
XM_017012381.1 | XP_016867870.1 | 13 - 14 | 1904 | 71 | |
XM_017012382.1 | XP_016867871.1 | 13 - 14 | 1873 | 71 | |
XM_017012383.1 | XP_016867872.1 | 13 - 14 | 1934 | 71 | |
XM_017012384.1 | XP_016867873.1 | 10 - 11 | 1441 | 71 | |
XM_017012385.1 | XP_016867874.1 | 12 - 13 | 1615 | 71 | |
XM_017012386.1 | XP_016867875.1 | 10 - 11 | 1394 | 71 | |
XM_017012387.1 | XP_016867876.1 | 13 - 14 | 1734 | 71 | |
XM_017012388.1 | XP_016867877.1 | 3 - 4 | 618 | 71 | |
GenBank mRNA | AK027228.1 | - | 2 - 3 | 215 | 71 |
AL137660.1 | - | 9 - 10 | 1031 | 71 | |
BC014491.1 | - | 13 - 14 | 1858 | 71 | |
CR627036.1 | - | 8 - 9 | 3500 | 71 |