Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 23 - 24 | 2937 | 120 |
XM_005249549.1 | XP_005249606.1 | 20 - 21 | 2529 | 120 | |
XM_006716041.2 | XP_006716104.1 | 23 - 24 | 2961 | 120 | |
XM_011516367.1 | XP_011514669.1 | 23 - 24 | 2803 | 120 | |
XM_011516369.2 | XP_011514671.1 | 23 - 24 | 2792 | 120 | |
XM_017012381.1 | XP_016867870.1 | 23 - 24 | 2983 | 120 | |
XM_017012383.1 | XP_016867872.1 | 23 - 24 | 3013 | 120 | |
XM_017012384.1 | XP_016867873.1 | 20 - 21 | 2520 | 120 | |
XM_017012385.1 | XP_016867874.1 | 22 - 23 | 2694 | 120 | |
XM_017012386.1 | XP_016867875.1 | 20 - 21 | 2473 | 120 | |
XM_017012387.1 | XP_016867876.1 | 23 - 24 | 2813 | 120 | |
XM_017012388.1 | XP_016867877.1 | 13 - 14 | 1697 | 120 | |
GenBank mRNA | AK001162.1 | - | 7 - 8 | 652 | 120 |
AK027228.1 | - | 12 - 13 | 1293 | 120 | |
AL137660.1 | - | 18 - 19 | 1937 | 120 | |
BC014491.1 | - | 23 - 24 | 2937 | 120 | |
CR627036.1 | - | 18 - 19 | 4579 | 120 |