Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290135.1 | NP_001277064.1 | 5 - 6 | 531 | 70 |
NM_004879.4 | NP_004870.3 | 5 - 6 | 573 | 70 | |
NR_110769.1 | 4 - 5 | 427 | 70 | ||
XM_011543069.1 | XP_011541371.1 | 6 - 7 | 589 | 70 | |
XM_011543070.1 | XP_011541372.1 | 5 - 6 | 744 | 70 | |
XM_017018582.1 | XP_016874071.1 | 6 - 7 | 589 | 70 | |
GenBank mRNA | AB209820.1 | - | 5 - 6 | 530 | 70 |
AF010313.2 | - | 5 - 6 | 445 | 70 | |
AK223029.1 | - | 5 - 6 | 502 | 70 | |
AK291951.1 | - | 5 - 6 | 482 | 70 | |
AK296620.1 | - | 5 - 6 | 460 | 70 | |
AK301556.1 | - | 4 - 5 | 356 | 70 | |
AK315841.1 | - | 5 - 6 | 489 | 70 | |
BC002390.2 | - | 5 - 6 | 439 | 70 |