Assay Design



Gene Transcripts

Gene Symbol
THUMPD2
Entrez Gene ID
80745
Gene Name
THUMP domain containing 2
Gene Aliases
C2orf8
Chromosome Location
Chr.2: 39736060 - 39779276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028102.1 7 - 8 1052 109
NR_135659.1 8 - 9 1343 109
NR_135660.1 8 - 9 1245 109
NR_135661.1 8 - 9 1333 109
NR_135662.1 7 - 8 1042 109
NR_135663.1 6 - 7 642 109
NR_135664.1 8 - 9 1235 109
GenBank mRNA AF380577.1 - 4 - 5 348 109
AF380578.1 - 4 - 5 1856 109
AF380579.1 - 4 - 5 1022 109
AK057801.1 - 5 - 6 434 109
AK093580.1 - 7 - 8 1023 109
AK292002.1 - 7 - 8 1037 109
AL832115.1 - 7 - 8 1019 109

View Full Product Details