Assay Design
Gene Transcripts
- Gene Symbol
- SNHG3
- Entrez Gene ID
-
8420
- Gene Name
- small nucleolar RNA host gene 3
- Gene Aliases
-
NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
- Chromosome Location
-
Chr.1: 28505943 - 28510892 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002909.2 |
|
2 - 3 |
176 |
132 |
NR_036473.1 |
|
3 - 4 |
284 |
240 |
GenBank mRNA |
DA044266.1 |
- |
2 - 3 |
135 |
132 |
DA135739.1 |
- |
3 - 4 |
247 |
240 |
- Gene Symbol
- RCC1
- Entrez Gene ID
-
1104
- Gene Name
- regulator of chromosome condensation 1
- Gene Aliases
-
CHC1, RCC1-I, SNHG3-RCC1
- Chromosome Location
-
Chr.1: 28505943 - 28539196 on Build GRCh38
View Full Product Details