Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083.3 | NP_001074.2 | 8 - 9 | 1466 | 148 |
NM_033430.2 | NP_236914.2 | 8 - 9 | 1297 | 148 | |
NM_033437.3 | NP_246273.2 | 8 - 9 | 1249 | 148 | |
XM_017008791.1 | XP_016864280.1 | 8 - 9 | 1720 | 148 | |
GenBank mRNA | AB015656.1 | - | 8 - 9 | 1191 | 148 |
AF043731.1 | - | 8 - 9 | 1466 | 148 | |
AJ004865.1 | - | 8 - 9 | 1404 | 148 | |
AK290189.1 | - | 8 - 9 | 1347 | 148 | |
AY264918.1 | - | 8 - 9 | 1185 | 148 | |
BC126233.1 | - | 8 - 9 | 1400 | 148 | |
JQ286346.1 | - | 8 - 9 | 1311 | 148 |