Assay Design
Gene Transcripts
- Gene Symbol
- PDE5A
- Entrez Gene ID
-
8654
- Gene Name
- phosphodiesterase 5A
- Gene Aliases
-
CGB-PDE, CN5A, PDE5
- Chromosome Location
-
Chr.4: 119494395 - 119629080 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_033437.3 |
NP_246273.2 |
1 - 2 |
89 |
64 |
GenBank mRNA |
CD511199.1 |
- |
1 - 2 |
54 |
64 |
View Full Product Details