Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142883.1 | NP_001136355.1 | 3 - 4 | 739 | 103 |
NM_054111.4 | NP_473452.2 | 2 - 3 | 610 | 103 | |
XM_005248842.3 | XP_005248899.1 | 3 - 4 | 519 | 103 | |
XM_005248843.3 | XP_005248900.1 | 1 - 2 | 380 | 103 | |
XM_011514295.2 | XP_011512597.1 | 2 - 3 | 622 | 103 | |
GenBank mRNA | AF393812.1 | - | 1 - 2 | 305 | 103 |
AK056586.2 | - | 2 - 3 | 530 | 103 | |
AK223572.1 | - | 2 - 3 | 533 | 103 | |
AK289935.1 | - | 3 - 4 | 659 | 103 | |
BC089389.1 | - | 2 - 3 | 610 | 103 |