Assay Design
Gene Transcripts
- Gene Symbol
- NXF2
- Entrez Gene ID
-
56001
- Gene Name
- nuclear RNA export factor 2
- Gene Aliases
-
CT39, TAPL-2, TCP11X2
- Chromosome Location
-
Chr.X: 102247100 - 102326720 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK303069.1 |
- |
7 - 8 |
912 |
104 |
View Full Product Details