Assay Design



Gene Transcripts

Gene Symbol
UBXN2A
Entrez Gene ID
165324
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
Chromosome Location
Chr.2: 23927201 - 24001000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011532633.2 XP_011530935.1 2 - 3 534 119
GenBank mRNA AK291192.1 - 2 - 3 456 119

View Full Product Details