Assay Design



Gene Transcripts

Gene Symbol
LRRC14
Entrez Gene ID
9684
Gene Name
leucine rich repeat containing 14
Gene Aliases
LRRC14A
Chromosome Location
Chr.8: 144517965 - 144525178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272036.1 NP_001258965.1 2 - 3 144 103
GenBank mRNA BC011377.1 - 2 - 3 104 103

View Full Product Details